Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMIGD3
Entrez Gene ID
104564307
Gene Name
transmembrane and immunoglobulin domain containing 3
Gene Aliases
-
Chromosome Location
Chr.3: 31745649 - 31829059 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079645.3 NP_001073113.2 2 - 3 200 144
GenBank mRNA BC109599.1 - 2 - 3 187 144
DT839214.1 - 2 - 3 172 144
DT846942.1 - 2 - 3 201 144

View Full Product Details