Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NFYB
Entrez Gene ID
614382
Gene Name
nuclear transcription factor Y subunit beta
Gene Aliases
-
Chromosome Location
Chr.5: 67767799 - 67787372 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079786.2 NP_001073254.1 1 - 1 570 91
GenBank mRNA BC109900.1 - 1 - 1 571 91

View Full Product Details