Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COQ2
Entrez Gene ID
504633
Gene Name
coenzyme Q2, polyprenyltransferase
Gene Aliases
PHB:PPT
Chromosome Location
Chr.6: 98037925 - 98065079 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045893.2 NP_001039358.1 4 - 5 675 79
GenBank mRNA BC112551.1 - 4 - 5 676 79

View Full Product Details