Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPHL
Entrez Gene ID
505457
Gene Name
biphenyl hydrolase like
Gene Aliases
-
Chromosome Location
Chr.23: 50529752 - 50551563 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045918.1 NP_001039383.1 2 - 3 231 134
GenBank mRNA BC112470.1 - 2 - 3 231 134

View Full Product Details