Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM4SF5
Entrez Gene ID
511869
Gene Name
transmembrane 4 L six family member 5
Gene Aliases
-
Chromosome Location
Chr.19: 26576895 - 26583222 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046102.2 NP_001039567.1 2 - 3 290 116
GenBank mRNA BC112643.1 - 2 - 3 291 116

View Full Product Details