Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF2S3
Entrez Gene ID
512350
Gene Name
eukaryotic translation initiation factor 2 subunit gamma
Gene Aliases
-
Chromosome Location
Chr.X: 119205850 - 119220743 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046117.2 NP_001039582.1 9 - 10 1019 92
GenBank mRNA BC112875.1 - 9 - 10 1020 92
BT030491.1 - 9 - 10 1004 92

View Full Product Details