Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TVP23B
Entrez Gene ID
513550
Gene Name
trans-golgi network vesicle protein 23 homolog B
Gene Aliases
FAM18B, FAM18B1, FAM18B2
Chromosome Location
Chr.19: 32983940 - 32993498 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046152.2 NP_001039617.1 3 - 4 343 94
GenBank mRNA BC113231.1 - 3 - 4 239 94

View Full Product Details