Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP6
Entrez Gene ID
515310
Gene Name
dual specificity phosphatase 6
Gene Aliases
-
Chromosome Location
Chr.5: 19212969 - 19217523 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046195.1 NP_001039660.1 1 - 2 856 55
GenBank mRNA BC105538.1 - 1 - 2 857 55
BT025424.1 - 1 - 2 856 55

View Full Product Details