Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTRF1L
Entrez Gene ID
534354
Gene Name
mitochondrial translational release factor 1 like
Gene Aliases
-
Chromosome Location
Chr.9: 89903839 - 89918654 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046378.1 NP_001039843.1 1 - 2 296 89
GenBank mRNA BC112806.1 - 1 - 2 296 89

View Full Product Details