Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PQBP1
Entrez Gene ID
534894
Gene Name
polyglutamine binding protein 1
Gene Aliases
-
Chromosome Location
Chr.X: 87013764 - 87019563 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046389.1 NP_001039854.1 4 - 5 1115 140
GenBank mRNA BC105573.1 - 4 - 5 1115 140

View Full Product Details