Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLPP
Entrez Gene ID
535981
Gene Name
caseinolytic mitochondrial matrix peptidase proteolytic subunit
Gene Aliases
-
Chromosome Location
Chr.7: 18079516 - 18084741 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046414.2 NP_001039879.1 2 - 3 301 68
GenBank mRNA BC112880.1 - 2 - 3 302 68

View Full Product Details