Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COQ8A
Entrez Gene ID
536925
Gene Name
coenzyme Q8A
Gene Aliases
ADCK3, CABC1
Chromosome Location
Chr.16: 29906092 - 29953577 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046419.2 NP_001039884.1 13 - 14 1673 74
GenBank mRNA BC114164.1 - 13 - 14 1674 74
BT026249.1 - 8 - 9 789 74

View Full Product Details