Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C22H3orf14
Entrez Gene ID
613635
Gene Name
chromosome 22 C3orf14 homolog
Gene Aliases
C3orf14
Chromosome Location
Chr.22: 38978148 - 38992728 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046510.1 NP_001039975.1 4 - 5 1032 121
GenBank mRNA BC113216.1 - 4 - 5 1032 121
DQ335871.1 - 3 - 4 270 121

View Full Product Details