Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DENR
Entrez Gene ID
614238
Gene Name
density regulated re-initiation and release factor
Gene Aliases
-
Chromosome Location
Chr.17: 52750180 - 52762092 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046528.1 NP_001039993.1 1 - 2 88 95
GenBank mRNA BC113316.1 - 1 - 2 88 95

View Full Product Details