Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
T2R12
Entrez Gene ID
664643
Gene Name
bitter taste receptor Bota-T2R12
Gene Aliases
-
Chromosome Location
Chr.5: 98661895 - 98662824 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046630.1 NP_001040095.1 1 - 1 849 73

View Full Product Details