Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPA1
Entrez Gene ID
280701
Gene Name
inorganic pyrophosphatase 1
Gene Aliases
PP
Chromosome Location
Chr.28: 26418192 - 26456501 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075118.1 NP_001068586.1 10 - 11 917 114
GenBank mRNA BC114891.1 - 10 - 11 917 114
M95283.1 - 10 - 11 903 114

View Full Product Details