Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP2B6
Entrez Gene ID
504769
Gene Name
cytochrome P450 subfamily 2B
Gene Aliases
CYP2B
Chromosome Location
Chr.18: 50290689 - 50308034 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075173.1 NP_001068641.1 7 - 8 1018 61
GenBank mRNA BC112659.1 - 7 - 8 1018 61
DQ087595.1 - 3 - 4 479 61

View Full Product Details