Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRAGB
Entrez Gene ID
506754
Gene Name
Ras related GTP binding B
Gene Aliases
-
Chromosome Location
Chr.X: 93035876 - 93077046 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075279.1 NP_001068747.1 1 - 2 146 108
GenBank mRNA BC118272.1 - 1 - 2 146 108

View Full Product Details