Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX15
Entrez Gene ID
507751
Gene Name
sorting nexin 15
Gene Aliases
-
Chromosome Location
Chr.29: 43242568 - 43254074 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075330.2 NP_001068798.1 2 - 3 187 70
GenBank mRNA BC118397.1 - 2 - 3 189 70

View Full Product Details