Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT79
Entrez Gene ID
508202
Gene Name
keratin 79
Gene Aliases
-
Chromosome Location
Chr.5: 27098154 - 27109004 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075348.1 NP_001068816.1 5 - 6 1052 115
GenBank mRNA BC118314.1 - 5 - 6 1052 115

View Full Product Details