Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUTA
Entrez Gene ID
508956
Gene Name
cutA divalent cation tolerance homolog
Gene Aliases
-
Chromosome Location
Chr.23: 7592050 - 7593652 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075379.1 NP_001068847.1 1 - 2 420 74
GenBank mRNA BC114795.1 - 1 - 2 420 74

View Full Product Details