Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HARBI1
Entrez Gene ID
514442
Gene Name
harbinger transposase derived 1
Gene Aliases
C11orf77, C15H11orf77
Chromosome Location
Chr.15: 76568903 - 76577894 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075668.1 NP_001069136.1 1 - 2 765 61
GenBank mRNA BC118217.1 - 1 - 2 765 61

View Full Product Details