Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSPAN5
Entrez Gene ID
538668
Gene Name
tetraspanin 5
Gene Aliases
-
Chromosome Location
Chr.6: 25923695 - 26102500 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076119.1 NP_001069587.1 6 - 7 982 119
GenBank mRNA BC118327.1 - 6 - 7 982 119
BC120462.1 - 3 - 4 321 119

View Full Product Details