Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRFN3
Entrez Gene ID
767839
Gene Name
leucine rich repeat and fibronectin type III domain containing 3
Gene Aliases
-
Chromosome Location
Chr.18: 46629617 - 46637139 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076959.1 NP_001070427.1 2 - 3 1838 71
GenBank mRNA BC114740.1 - 2 - 3 1838 71

View Full Product Details