Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT2
Entrez Gene ID
768044
Gene Name
nudix hydrolase 2
Gene Aliases
-
Chromosome Location
Chr.8: 75724352 - 75735276 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077102.1 NP_001070570.1 2 - 3 214 82
GenBank mRNA BC114149.1 - 2 - 3 214 82

View Full Product Details