Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DHFR
Entrez Gene ID
508809
Gene Name
dihydrofolate reductase
Gene Aliases
-
Chromosome Location
Chr.7: 80733288 - 80748878 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077883.1 NP_001071351.1 1 - 2 162 125
GenBank mRNA BC114162.1 - 1 - 2 162 125

View Full Product Details