Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HCRTR1
Entrez Gene ID
282248
Gene Name
hypocretin receptor 1
Gene Aliases
-
Chromosome Location
Chr.2: 122098185 - 122108098 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048182.1 NP_001041647.1 6 - 7 1090 77
GenBank mRNA BC149920.1 - 8 - 9 1794 77
DQ874350.1 - 6 - 7 1090 77

View Full Product Details