Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD10
Entrez Gene ID
505888
Gene Name
BTB domain containing 10
Gene Aliases
-
Chromosome Location
Chr.15: 39210355 - 39285718 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075230.1 NP_001068698.1 1 - 2 235 69
GenBank mRNA BC119974.1 - 1 - 2 235 69

View Full Product Details