Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC3
Entrez Gene ID
506728
Gene Name
zinc finger DHHC-type palmitoyltransferase 3
Gene Aliases
-
Chromosome Location
Chr.22: 54204466 - 54264043 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075278.1 NP_001068746.1 3 - 4 707 97
GenBank mRNA BC123521.1 - 3 - 4 707 97

View Full Product Details