Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANP32E
Entrez Gene ID
507203
Gene Name
acidic nuclear phosphoprotein 32 family member E
Gene Aliases
-
Chromosome Location
Chr.3: 20409583 - 20421686 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075306.1 NP_001068774.1 2 - 3 507 72
GenBank mRNA BC120174.1 - 2 - 3 507 72

View Full Product Details