Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP3
Entrez Gene ID
508138
Gene Name
THAP domain containing 3
Gene Aliases
-
Chromosome Location
Chr.16: 46777982 - 46783605 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075347.1 NP_001068815.1 3 - 4 336 107
GenBank mRNA BC120270.1 - 3 - 4 336 107

View Full Product Details