Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG3
Entrez Gene ID
508571
Gene Name
autophagy related 3
Gene Aliases
-
Chromosome Location
Chr.1: 57309112 - 57343709 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075364.1 NP_001068832.1 3 - 4 583 74
GenBank mRNA BC120110.1 - 3 - 4 583 74

View Full Product Details