Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX10
Entrez Gene ID
508836
Gene Name
sorting nexin 10
Gene Aliases
-
Chromosome Location
Chr.4: 69587802 - 69655624 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075375.1 NP_001068843.1 6 - 7 666 104
GenBank mRNA BC122586.1 - 6 - 7 666 104

View Full Product Details