Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR4X5
Entrez Gene ID
510452
Gene Name
olfactory receptor family 4 subfamily X member 5
Gene Aliases
OR4X1
Chromosome Location
Chr.15: 77950829 - 77951758 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075450.1 NP_001068918.1 1 - 1 452 105

View Full Product Details