Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD39
Entrez Gene ID
510840
Gene Name
ankyrin repeat domain 39
Gene Aliases
-
Chromosome Location
Chr.11: 2807893 - 2815163 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075471.2 NP_001068939.1 1 - 2 129 62
GenBank mRNA BC120254.1 - 1 - 2 130 62

View Full Product Details