Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGD2
Entrez Gene ID
512593
Gene Name
FYVE, RhoGEF and PH domain containing 2
Gene Aliases
-
Chromosome Location
Chr.23: 10916192 - 10942801 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075570.1 NP_001069038.1 12 - 13 1487 146
GenBank mRNA BT026545.1 - 12 - 13 1487 146

View Full Product Details