Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCN2
Entrez Gene ID
512717
Gene Name
reticulocalbin 2
Gene Aliases
-
Chromosome Location
Chr.21: 32049173 - 32067326 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075579.1 NP_001069047.1 6 - 7 1026 82
GenBank mRNA BC120049.1 - 6 - 7 1026 82

View Full Product Details