The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075703.1 | NP_001069171.1 | 7 - 8 | 1344 | 81 |
GenBank mRNA | BT026535.1 | - | 7 - 8 | 1344 | 81 |