The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075846.2 | NP_001069314.1 | 3 - 4 | 286 | 60 |
GenBank mRNA | BC122841.1 | - | 3 - 4 | 287 | 60 |