Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP2CB
Entrez Gene ID
524361
Gene Name
protein phosphatase 2 catalytic subunit beta
Gene Aliases
-
Chromosome Location
Chr.27: 26900095 - 26923698 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075857.1 NP_001069325.1 6 - 7 863 71
GenBank mRNA BC120332.1 - 6 - 7 863 71

View Full Product Details