Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGPAT5
Entrez Gene ID
530414
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 5
Gene Aliases
-
Chromosome Location
Chr.27: 5810043 - 5857909 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075932.1 NP_001069400.1 6 - 7 876 136
GenBank mRNA BC122693.1 - 6 - 7 876 136

View Full Product Details