Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NMRAL1
Entrez Gene ID
534628
Gene Name
NmrA like redox sensor 1
Gene Aliases
-
Chromosome Location
Chr.25: 3608220 - 3614429 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076032.2 NP_001069500.1 1 - 2 42 64
GenBank mRNA BC120089.1 - 1 - 2 43 64

View Full Product Details