Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTI1
Entrez Gene ID
534917
Gene Name
TELO2 interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.13: 66868169 - 66909599 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076042.1 NP_001069510.1 3 - 4 2708 75
GenBank mRNA BC126508.1 - 4 - 5 2720 75
BT026193.1 - 3 - 4 2708 75

View Full Product Details