Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TWSG1
Entrez Gene ID
537290
Gene Name
twisted gastrulation BMP signaling modulator 1
Gene Aliases
-
Chromosome Location
Chr.24: 41546086 - 41567761 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076089.1 NP_001069557.1 3 - 4 375 64
GenBank mRNA BC119840.1 - 3 - 4 375 64

View Full Product Details