Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALG10
Entrez Gene ID
539837
Gene Name
ALG10, alpha-1,2-glucosyltransferase
Gene Aliases
ALG10B
Chromosome Location
Chr.5: 76106754 - 76113238 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076189.1 NP_001069657.2 1 - 2 292 70
GenBank mRNA BC119908.1 - 1 - 2 292 70
BT030571.1 - 1 - 2 252 70

View Full Product Details