Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICE2
Entrez Gene ID
541255
Gene Name
interactor of little elongation complex ELL subunit 2
Gene Aliases
NARG2
Chromosome Location
Chr.10: 49706932 - 49772346 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076265.1 NP_001069733.1 14 - 15 2811 139
GenBank mRNA BC120051.1 - 14 - 15 2811 139

View Full Product Details