Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAS4
Entrez Gene ID
617710
Gene Name
breast carcinoma amplified sequence 4
Gene Aliases
-
Chromosome Location
Chr.13: 78752441 - 78808954 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076474.2 NP_001069942.1 4 - 5 455 90
GenBank mRNA BC122730.1 - 4 - 5 456 90

View Full Product Details