Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RABL6
Entrez Gene ID
508218
Gene Name
RAB, member RAS oncogene family like 6
Gene Aliases
C11H9orf86, PARF, RBEL1
Chromosome Location
Chr.11: 105908984 - 105928079 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076817.1 NP_001070285.1 14 - 15 2079 122
GenBank mRNA BC123865.1 - 14 - 15 2079 122

View Full Product Details