Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CDH18
Entrez Gene ID
512080
Gene Name
cadherin 18
Gene Aliases
-
Chromosome Location
Chr.20: 53333336 - 53938648 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076837.1 NP_001070305.1 7 - 8 1560 95
GenBank mRNA BC123714.1 - 7 - 8 1560 95

View Full Product Details