Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM267
Entrez Gene ID
780867
Gene Name
transmembrane protein 267
Gene Aliases
C20H5orf28, C5orf28
Chromosome Location
Chr.20: 31332643 - 31353180 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079795.1 NP_001073263.1 2 - 3 137 124
GenBank mRNA BC118330.1 - 2 - 3 179 124
BT026226.1 - 2 - 3 137 124

View Full Product Details